Human Bub3 peptide (16 aa near C-terminus)
PRSI4227P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near C-terminus
- Protein/peptide name:Bub3 Peptide (16 aa near C-terminus)
- Tested applications:Blocking
Bub3 peptide is used for blocking the activity of Bub3 antibody.