Human PDL-2 peptide (16 aa in the center)
Catalog # PRSI4063P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa in the center
- Protein/peptide name:PDL-2 Peptide (16 aa in the center)
- Tested applications:Blocking
Specifications
About this item
The peptide is used for blocking the activity of PDL-2 antibody.