Human PDL-1 peptide (17 aa in the center)
PRSI4059P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa in the center
- Protein/peptide name:PDL-1 Peptide (17 aa in the center)
- Tested applications:Blocking
The peptide is used for blocking the activity of PDL-1 antibody.