Human TSLP peptide (19 aa in the center)
PRSI4023P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa in the center
- Protein/peptide name:TSLP Peptide (19 aa in the center)
- Tested applications:Blocking
TSLP peptide is used for blocking the activity of TSLP antibody.