Order Entry
Export
ContactUsLinkComponent
West nile virus WNV envelope peptide (15 aa near the center)
West nile virus WNV envelope peptide (15 aa near the center)
Catalog # PRSI3437P
Supplier:  ProSci Inc.
CAS Number:  
West nile virus WNV envelope peptide (15 aa near the center)
Catalog # PRSI3437P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    West nile virus
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near the center
  • Protein/peptide name:
    WNV Envelope Peptide (15 aa near the center)
  • Tested applications:
    Blocking

Specifications

About this item

West Nile virus peptide is used for blocking the activity of West Nile virus envelope antibody.