Human ACE2 peptide (17 aa in the center)
PRSI3215P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa in the center
- Protein synonyms:Angiotensin Converting Enzyme 2
- Protein/peptide name:ACE2 Peptide (17 aa in the center)
- Tested applications:Blocking
ACE2 peptide is used for blocking the activity of ACE2 antibody.