Mouse Caspase 12 (18 aa) peptide
PRSI3195P
:
- Pk:50 µG
- Species:Mouse
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:Caspase 12 (18 aa) Peptide
- Tested applications:Blocking
Caspase-12 peptide is used for blocking the activity of the Caspase-12 antibody.