Human PID (17 aa) peptide
Catalog # PRSI2443P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:PID (17 aa) Peptide
- Tested applications:Blocking
Specifications
About this item
PID peptide is used for blocking the activity of PID antibody.