Order Entry
Export
ContactUsLinkComponent
Human Bnip3L (16 aa) peptide
Human Bnip3L (16 aa) peptide
Catalog # PRSI2289P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Bnip3L (16 aa) peptide
Catalog # PRSI2289P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Protein/peptide name:
    Bnip3L (16 aa) Peptide
  • Tested applications:
    Blocking

Specifications

About this item

Bnip3L peptide is used for blocking the activity of Bnip3L antibody.