Order Entry
Export
ContactUsLinkComponent
 
Human DR4 peptide (19 aa near C-terminus)
  PRSI1139P
 :  
Human DR4 peptide (19 aa near C-terminus)
  PRSI1139P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa near C-terminus
  • Protein/peptide name:
    DR4 Peptide (19 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

DR4 peptide is used for blocking the activity of DR4 antibody.