Human DcR1 peptide (19 aa near the extracellular domain)
Catalog # PRSI2179P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near the extracellular domain
- Protein/peptide name:DcR1 Peptide (19 aa near the extracellular domain)
- Tested applications:Blocking
Specifications
About this item
DcR1 peptide is used for blocking the activity of DcR1 antibody.