Anti-CCL25 Mouse Monoclonal Antibody [clone: 1.2_4G1-1G4-1C9]
PRSIXP-5725-M
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 25
- Antigen symbol:CCL25
- Clonality:Monoclonal
- Clone:1.2_4G1-1G4-1C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:6370
- Antigen synonyms:CCL25|Chemokine TECKTECK|Small inducible cytokine A25|C-C motif chemokine 25|TECK|SCYA25|Thymus expressed chemokine|Ckb15
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in Balb/c X ICR F1 mice using recombinant hTECK as the immunizing antigen.
- Purification:This IgG1k antibody was purified from ascites fluid on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
This gene belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. TECK displays chemotactic activity for dendritic cells, thymocytes, and activated macrophages but is inactive on peripheral blood lymphocytes and neutrophils. TECK binds to chemokine receptor CCR9. Two alternatively spliced variants encoding two different isoforms have been identified.
Type: Primary
Antigen: CCL25
Clonality: Monoclonal
Clone: 1.2_4G1-1G4-1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human