Anti-NOS1 Rabbit Polyclonal Antibody
Catalog # PRSI80-024
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Nitric Oxide Synthase 1
- Antigen symbol:NOS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4842
- Antigen synonyms:N-NOS|Peptidyl-cysteine S-nitrosylase NOS1|Constitutive NOS|NC-NOS|Nitric oxide synthase|bNOS|brain|NOS1|NOS type I|nNOS|Neuronal NOS
- Modification:phospho-Ser852
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:120, 160 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:-20°C
- Immunogen:n-NOS (phospho Ser852) antibody was raised against a peptide sequence around phosphorylation site of serine 852 (F-N-S (p)-V-S) derived from Human n-NOS .
- Purification:Purified by affinity-chromatography using epitope-specific peptide.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Produces nitric oxide (NO) which is a messenger molecule with diverse functions throughout the body. In the brain and peripheral nervous system, NO displays many properties of a neurotransmitter. Probably has nitrosylase activity and mediates cysteine S-nitrosylation of cytoplasmic target proteins such SRR.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: NOS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat