Order Entry
Export
ContactUsLinkComponent
Anti-SMAD3 Rabbit Polyclonal Antibody
Anti-SMAD3 Rabbit Polyclonal Antibody
Catalog # PRSI80-005
Supplier:  ProSci Inc.
undefined
Anti-SMAD3 Rabbit Polyclonal Antibody
Catalog # PRSI80-005
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SMAD Family Member 3
  • Antigen symbol:
    SMAD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    4088
  • Antigen synonyms:
    DKFZp586N0721|JV15-2|DKFZp686J10186|Mad3 |Mothers against decapentaplegic homolog 3|HsT17436|hSMAD3|hMAD-3|HSPC193|MGC60396|MADH3
  • Modification:
    phospho-Thr179
  • Storage buffer:
    Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
  • Molecular weight:
    50 kDa
  • Storage temperature:
    Store antibody at –20 °C for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    -20°C
  • Immunogen:
    Smad3 (phospho Thr179) antibody was raised against a peptide sequence around phosphorylation site of threonine179 (P-E-T (p)-P-P) derived from Human Smad3 .
  • Purification:
    Purified by affinity-chromatography using epitope-specific peptide.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Smad3 encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.

Western Blot: 1:500~1:1000

Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat