Anti-Ccl12 Rabbit Polyclonal Antibody
PRSIXP-5226
- Antigen symbol:Ccl12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mMCP-5 (murine MCP-5).
- Size:100 µg
- Pk:100 µG
MCP-5 is a member of the b-chemokine family (C-C) of cytokines. This chemokine is strongly chemotactic for human monocytes and murine peritoneal macrophages, but is not active on mouse neutrophils and shows only a weak response on mouse eosinophils at high concentrations
Anti-mMCP-5 specific antibody was purified by affinity chromatography employing immobilized mMCP-5 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Ccl12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse