Specifications
- Antigen name:Lin-28 homolog A
- Antigen symbol:LIN28
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RNA-binding proteináLIN-28|Zinc finger CCHC domain-containing protein 1|Lin-28 homolog A (C. elegans)|CSDD1|ZCCHC1|LIN28A
- Concentration:1 mg/mL
- Immunogen:Mouse monoclonal Lin28 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human Lin28.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Lin28 Monoclonal Antibody: Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
Lin28 antibody can be used for detection of Lin28 by Western blot at 0.5 - 1 μg/mL.
Lin28 Monoclonal Antibody is affinity chromatography purified via peptide column.
Lin28 Monoclonal Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LIN28
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human