Anti-SS18 Rabbit Polyclonal Antibody
Catalog # PRSI79-655
Supplier: ProSci Inc.
Specifications
- Antigen name:Synovial Sarcoma Translocation, Chromosome 18
- Antigen symbol:SS18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:SSXT|synovial sarcoma translocation|SYT|chromosome 18
- Concentration:1 mg/mL
- Immunogen:SYT antibody was raised against a peptide sequence around aa.14~18 (G-E-I-T-P) derived from Human SYT.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Appears to function synergistically with RBM14 as a transcriptional coactivator. Isoform 1 and isoform 2 function in nuclear receptor coactivation. Isoform 1 and isoform 2 function in general transcriptional coactivation.
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SS18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat