- Antigen name:T-box 21
- Antigen symbol:TBX21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:T-box transcription factor TBX21|TBX21 |T-cell-specific T-box transcription factor T-bet|T-PET|T-box protein 21|T-bet|TBLYM|TBET
- Concentration:1 mg/mL
- Immunogen:TBX21 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human TBX2.
- Size:100 µg
- Pk:100 µG
TBX21 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box (1,2). Members of this family include transcription factors involved in the regulation of developmental processes. Studies in mouse and humans show that TBX21 is a Th1 cell-specific transcription factor that controls the expression of the hallmark Th1 cytokine, interferon-gamma (1,3). Expression of the human ortholog also correlates with interferon-gamma expression in Th1 and natural killer cells, suggesting a TBX21 may play a role in initiating Th1 lineage development from naive Th precursor cells..
TBX21 Antibody is affinity chromatography purified via peptide column.
TBX21 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TBX21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse