- Antigen name:Tripartite Motif Containing 33
- Antigen symbol:TRIM33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TIF1GAMMA|KIAA1113|TIFGAMMA|TF1G|E3 ubiquitin-protein ligase TRIM33|Protein Rfg7|TIF1G|RFG7|PTC7|Ectodermin homolog|ECTO|TRIM33
- Concentration:1 mg/mL
- Immunogen:TRIM33 antibody was raised against an 18 amino acid peptide near the center of human TRIM33.
- Size:100 µg
- Pk:100 µG
TRIM33 Antibody: TRIM24 (TIF1alpha), TRIM28 (TIF1beta), and TRIM33 (TIF1gamma) are three related cofactors belonging to the tripartite motif superfamily that interact with distinct transcription factors. The structure of TRIM33 is similar to TRIM24 and TRIM28, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TRIM33 functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells. It interacts with the Smad2/3 transcription factor in hematopoietic, mesenchymal, and epithelial cell types to mediate different transcriptional effects in response to TGF-beta. It acts as an E3 ubiquitin-protein ligase and promotes SMAD4 ubiquitination.
TRIM33 Antibody is affinity chromatography purified via peptide column.
TRIM33 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat