Specifications
- Antigen name:Parkin
- Antigen symbol:PARK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Parkinson disease protein 2|PARK2|PRKN|Parkinson juvenile disease protein 2|Ubiquitin E3 ligase PRKN
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser378 of human parkin.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Parkin is an E3 ligase in the ubiquitin-proteasome system. Hereditary Parkinson’s disease is most commonly caused by mutations in the parkin gene and is characterized by the progressive loss of dopaminergic neurons and the presence of Lewy bodies in the substania nigra (Jenner aet al.,1992). Recent evidence suggests that phosphorylation of parkin at Ser378 may have an important regulatory role on its E3 ubiquitin ligase activity (Yamamoto et al., 2005).
The antibody has been directly tested for reactivity in Western blots with human tissue.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PARK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human