Anti-CCNE1 Rabbit Polyclonal Antibody
Catalog # PRSI47-135
Supplier: ProSci Inc.
Specifications
- Antigen name:Cyclin E1
- Antigen symbol:CCNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
- Immunogen:Synthetic peptide corresponding to aa residues 515-535 of mouse SHPRH conjugated to KLH at the N-terminus.
- Tested applications:Immunoaffinity Chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus, suggests a role for SHPRH in tumor suppression.
PBS
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CCNE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse