Anti-TXNDC4 Goat Polyclonal Antibody
Catalog # PRSI46-542
Supplier: ProSci Inc.
Specifications
- Antigen name:Thioredoxin Domain Containing 4
- Antigen symbol:TXNDC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:OTTHUMP00000063799|KIAA0573|TXNDC4 |thioredoxin domain containing 4 (endoplasmic reticulum) |endoplasmic reticulum resident protein 44 kDa|ERP44
- Concentration:500 ug/mL
- Immunogen:TXNDC4 antibody was raised against a 13 amino acid synthetic peptide near the internal region of TXNDC4.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TXNDC4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
TXNDC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TXNDC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human