Anti-RHBG Goat Polyclonal Antibody
Catalog # PRSI42-643
Supplier: ProSci Inc.
Specifications
- Antigen name:Rh Family B glycoprotein (gene/pseudogene)
- Antigen symbol:RHBG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:RHBG antibody was raised against a 10 amino acid synthetic peptide near the C-Terminus of RHBG (mouse).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
RHBG antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
RHBG antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RHBG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse