Anti-SPINT2 Goat Polyclonal Antibody
PRSI42-634
- Antigen name:Serine Peptidase Inhibitor, Kunitz Type 2
- Antigen symbol:SPINT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:placental bikunin|Kunitz type|hepatocyte growth factor activator inhibitor type 2|serine protease inhibitor|FLJ45571|HAI2|2|Kop|PB|HAI-2|DIAR3
- Concentration:500 ug/mL
- Immunogen:SPINT2 antibody was raised against a 12 amino acid synthetic peptide near the internal region of SPINT2.
- Size:100 µg
- Pk:100 µG
SPINT2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
SPINT2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SPINT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human