Order Entry
Export
ContactUsLinkComponent
Anti-POU2F3 Goat Polyclonal Antibody
Anti-POU2F3 Goat Polyclonal Antibody
Catalog # PRSI42-576
Supplier:  ProSci Inc.
undefined
Anti-POU2F3 Goat Polyclonal Antibody
Catalog # PRSI42-576
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    POU Class 2 Homeobox 3
  • Antigen symbol:
    POU2F3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    POU transcription factor|MGC126698|Skn-1a|class 2|Epoc-1|OCT11|POU class 2 homeobox 3|PLA-1|transcription factor 3|POU domain|POU2F3|FLJ40063
  • Concentration:
    500 ug/mL
  • Immunogen:
    POU2F3 antibody was raised against a 15 amino acid synthetic peptide near the internal region of POU2F3.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

POU2F3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

POU2F3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POU2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human