Anti-HTR7 Rabbit Polyclonal Antibody
PRSI31-118
- Antigen name:5-Hydroxytryptamine (serotonin) Receptor 7 (adenylate cyclase-coupled)
- Antigen symbol:HTR7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Serotonin receptor 7|5-HT7,5-HT-X
- Immunogen:A synthetic peptide corresponding a region of human HTR7.
- Size:50 µg
- Pk:50 µG
This is one of the several different receptors for 5- hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by g proteins that stimulate adenylate cyclase.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HTR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human