Anti-HTR1A Rabbit Polyclonal Antibody
Catalog # PRSI31-113
Supplier: ProSci Inc.
Specifications
- Antigen name:5-Hydroxytryptamine (Serotonin) Receptor 1A
- Antigen symbol:HTR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ADRB2RL1|5-HT1A|ADRBRL1|5-HT-1A|PFMCD|5HT1a|G-21
- Immunogen:A synthetic peptide corresponding a region of human HTR1A.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This is one of the several different receptors for 5- hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibits adenylate cyclase activity.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HTR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human