Specifications
- Antigen name:Solute Carrier Family 25, Member 31
- Antigen symbol:SLC25A31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ADP/ATP translocase 4|Solute carrier family 25 member 31|ADP,ATP carrier protein 4|AAC4|Adenine nucleotide translocator 4|SLC25A31|Sperm flagellar energy carrier protein|ANT4|SFEC
- Immunogen:A synthetic peptide corresponding a region of human SLC25A31.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Mitochondrial ADP/ATP carriers, such as SLC25A31, are nuclear-coded mitochondrial proteins that catalyze the exchange of ATP generated in mitochondria by ATP synthase (see MIM 108729) against ADP produced in cytosol by most energy-consuming reactions.Mitochondrial ADP/ATP carriers, such as SLC25A31, are nuclear-coded mitochondrial proteins that catalyze the exchange of ATP generated in mitochondria by ATP synthase (see MIM 108729) against ADP produced in cytosol by most energy-consuming reactions (Dolce et al., 2005 [PubMed 15670820]).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC25A31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat