- Antigen name:ADAM Metallopeptidase Domain 33
- Antigen symbol:ADAM33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:disintegrin and reprolysin metalloproteinase family protein|MGC71889|a disintegrin and metalloprotease 33|DJ964F7.1|FLJ35308|a disintegrin and metalloproteinase domain 33|MGC149823|C20orf153|DKFZp434K0521|RP5-964F7.2|ADAM33|HGNC:15478|FLJ36751|metalloprotease disintegrin|ADAM metallopeptidase domain 33
- Immunogen:A synthetic peptide corresponding a region of human ADAM33.
- Size:50 µg
- Pk:50 µG
ADAM33 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM33 is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness.This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ADAM33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat