Specifications
- Antigen name:alpha 1,3/1,6-Mannosyltransferase
- Antigen symbol:ALG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ALG2
- Immunogen:A synthetic peptide corresponding a region of human ALG2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ALG2 is a member of the glycosyltransferase 1 family. It acts as an alpha 1,3 mannosyltransferase, mannosylating Man (2)GlcNAc (2)-dolichol diphosphate and Man (1)GlcNAc (2)-dolichol diphosphate to form Man (3)GlcNAc (2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).This gene encodes a member of the glycosyltransferase 1 family. The encoded protein acts as an alpha 1,3 mannosyltransferase, mannosylating Man (2)GlcNAc (2)-dolichol diphosphate and Man (1)GlcNAc (2)-dolichol diphosphate to form Man (3)GlcNAc (2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii). Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ALG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat