Anti-TAMM41 Rabbit Polyclonal Antibody
PRSI30-314
- Antigen name:TAM41 mitochondrial translocator assembly and maintenance homolog
- Antigen symbol:TAMM41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:C3orf31|TAM41|RAM41
- Immunogen:A synthetic peptide corresponding a region of human C3orf31.
- Size:100 µg
- Pk:100 µG
C3orf31 may be involved in the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TAMM41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human