Anti-SLC9A9 Rabbit Polyclonal Antibody
Catalog # PRSI29-957
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 9 Member 9
- Antigen symbol:SLC9A9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:5730527A11Rik|AI854429|9930105B05|Nhe9
- Immunogen:A synthetic peptide corresponding a region of human SLC9A9.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC9A9 may act in electroneutral exchange of protons for Na (+) across membranes.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC9A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse