Specifications
- Antigen name:Solute Carrier Family 39 Member A5
- Antigen symbol:SLC39A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Zinc transporter ZIP5|Solute carrier family 39 member 5|LZT-Hs7|ZIP5|ZIP-5
- Immunogen:A synthetic peptide corresponding a region of human SLC39A5.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A5 belongs to a subfamily of proteins that show structural characteristics of zinc transporters.Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A5 belongs to a subfamily of proteins that show structural characteristics of zinc transporters (Taylor and Nicholson, 2003).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC39A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat