Anti-SLCO6A1 Rabbit Polyclonal Antibody
PRSI29-954
- Antigen name:Solute Carrier Organic Anion Transporter Family member 6A1
- Antigen symbol:SLCO6A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:OATP-I|GST|CT48|OATPY|OATP6A1
- Immunogen:A synthetic peptide corresponding a region of human SLCO6A1.
- Size:100 µg
- Pk:100 µG
The function remains unknown.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLCO6A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human