Order Entry
Export
ContactUsLinkComponent
Anti-SLC22A2 Rabbit Polyclonal Antibody
Anti-SLC22A2 Rabbit Polyclonal Antibody
Catalog # PRSI29-915
Supplier:  ProSci Inc.
undefined
Anti-SLC22A2 Rabbit Polyclonal Antibody
Catalog # PRSI29-915
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 22 Member A2
  • Antigen symbol:
    SLC22A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    OCT2|hOCT2|Organic cation transporter 2|Solute carrier family 22 member 2
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC22A2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. SLC22A2 is one of the three similar cation transporters. SLC22A2 contains twelve putative transmembrane domains and is a plasma integral membrane protein. It is found primarily in the kidney, where it may mediate the first step in cation reabsorption.Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. It is found primarily in the kidney, where it may mediate the first step in cation reabsorption. Two transcript variants encoding different isoforms have been found for this gene.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC22A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human