Specifications
- Antigen name:Ring Finger Protein 39
- Antigen symbol:RNF39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:RNF39|HZFW1|LTP (long-term potentiation) induced RING finger protein|LIRF|HZF|HZFW|DADB-60O11.8|HZFw1 protein
- Immunogen:A synthetic peptide corresponding a region of human RNF39.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Its gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that RNF39 plays a role in an early phase of synaptic plasticity. Its gene lies within the major histocompatibility complex class I region on chromosome 6.This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Alternative splicing results in three transcript variants encoding different isoforms.This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Alternative splicing results in three transcript variants encoding different isoforms.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RNF39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human