Anti-Aly/REF export Factor Rabbit Polyclonal Antibody
Product Details & Documents
THOC4 is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Aly/REF export Factor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Documents
10109-042.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Form:Lyophilized
- Host:Rabbit
- Antigen name:Aly/REF export Factor
- Cat. No.:PRSI29-831
- Reactivity:Human|Rat|Mouse
- Antigen synonyms:ALYREF
- Immunogen:A synthetic peptide corresponding a region of human THOC4.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:Aly/REF export Factor
- ELISA:Yes



