- Antigen name:DEAD (Asp-Glu-Ala-Asp) box helicase 17
- Antigen symbol:DDX17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:DEAD/H box 17|RH70|RP3-434P1.1|DEAD box protein 17|RNA-dependent helicase p72|P72|DEAD box protein p72|probable ATP-dependent RNA helicase DDX17|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD)|DKFZp761H2016
- Immunogen:A synthetic peptide corresponding a region of human DDX17.
- Size:100 µg
- Pk:100 µG
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The DEAD box protein is an ATPase activated by a variety of RNA species but not by dsDNA. This protein and that encoded by DDX5 gene are more closely related to each other than to any other member of the DEAD box family.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DDX17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human