Order Entry
Export
ContactUsLinkComponent
Anti-SSB Rabbit Polyclonal Antibody
Anti-SSB Rabbit Polyclonal Antibody
Catalog # PRSI29-308
Supplier:  ProSci Inc.
undefined
Anti-SSB Rabbit Polyclonal Antibody
Catalog # PRSI29-308
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Single-strand DNA-binding protein
  • Antigen symbol:
    SSB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    La ribonucleoprotein|Sjoegren syndrome type B antigen|SS-B|La autoantigen|Lupus La protein
  • Immunogen:
    A synthetic peptide corresponding a region of human SSB.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

SSB is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU (OH) elements of newly RNA polymerase III-transcribed RNA, processing 5-prime and 3-prime ends of pre-tRNA precursors, acting as an RNA chaperone, and binding viral RNAs associated with hepatitis C virus. SSB protein was originally defined by its reactivity with autoantibodies from patients with Sjogren syndrome and systemic lupus erythematosus.La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU (OH) elements of newly RNA polymerase III (see MIM 606007)-transcribed RNA, processing 5-prime and 3-prime ends of pre-tRNA precursors, acting as an RNA chaperone, and binding viral RNAs associated with hepatitis C virus. La protein was originally defined by its reactivity with autoantibodies from patients with Sjogren syndrome (MIM 270150) and systemic lupus erythematosus La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU (OH) elements of newly RNA polymerase III (see MIM 606007)-transcribed RNA, processing 5-prime and 3-prime ends of pre-tRNA precursors, acting as an RNA chaperone, and binding viral RNAs associated with hepatitis C virus. La protein was originally defined by its reactivity with autoantibodies from patients with Sjogren syndrome (MIM 270150) and systemic lupus erythematosus (SLE; MIM 152700) (Teplova et al., 2006 [PubMed 16387655]).[supplied by OMIM]. Sequence Note: removed 1 base from the 3' end that did not align to the reference genome assembly. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-1673 BC020818.1 1-1673

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human