Anti-TRIM62 Rabbit Polyclonal Antibody
PRSI29-048
- Antigen name:Tripartite Motif Containing 62
- Antigen symbol:TRIM62
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human TRIM62.
- Size:100 µg
- Pk:100 µG
TRIM62 contains 1 RING-type zinc finger, which is probably involved in mediating protein-protein interactions. RING-type zinc finger was identified in a group of proteins with a wide range of functions such as viral replication, signal transduction, and development. But the function of TRIM62 remains unknown.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat