Order Entry
Export
ContactUsLinkComponent
 
Anti-TRIM16 Rabbit Polyclonal Antibody
Catalog # PRSI28-947
Supplier:  ProSci Inc.
undefined
Anti-TRIM16 Rabbit Polyclonal Antibody
Catalog # PRSI28-947
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Tripartite Motif Containing 16
  • Antigen symbol:
    TRIM16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    tripartite motif containing 16|EBBP
  • Immunogen:
    A synthetic peptide corresponding a region of human TRIM16.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. TRIM16 contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Its function, however, has not yet been determined.This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human