Specifications
- Antigen name:Jun B Transcription Factor
- Antigen symbol:JUNB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:AP-1|activator protein 1|Jun B Transcription Factor|Transcription factor jun-BGene Symbols: JUNB|JUNB
- Immunogen:A synthetic peptide corresponding a region of human JUNB.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The c-Jun proto-oncogene was first identified as the cellular homolog of the avian sarcoma virus v-Jun oncogene. The c-Jun protein, along with c-Fos, is a component of the AP-1 transcriptional complex. c-Jun can form either Jun/Jun homodimers or Jun/Fos heterodimers via the leucine repeats in both proteins. Jun B and Jun D, have been shown to be almost identical to c-Jun in their C-terminal regions, which are involved in dimerization and DNA binding, whereas their N-terminal domains, which are involved in transcriptional activation, diverge. JunB is involved in many types of human carcinoma including T-cell lymphomas, CML,primary cutaneous lymphomas. Aberrantly expressed c-Jun and JunB are a hallmark of Hodgkin lymphoma cells, stimulate proliferation and synergize with NF-kappa B. JunB potentiates function of BRCA1 activation domain 1 (AD1) through a coiled-coil-mediated interaction.JunB is an important regulator of erythroid Differentiation.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: JUNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human