Anti-DDX49 Rabbit Polyclonal Antibody
Catalog # PRSI28-497
Supplier: ProSci Inc.
Specifications
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box Polypeptide 49
- Antigen symbol:DDX49
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human DDX49.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The function of Anti-DDX49 has not yet been determined.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DDX49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse