Specifications
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box polypeptide 41
- Antigen symbol:DDX41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ABS|DEAD box protein 41|Probable ATP-dependent RNA helicase DDX41
- Immunogen:A synthetic peptide corresponding a region of human DDX41.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Based on studies in Drosophila, the abstrakt gene is widely required during post-transcriptional gene expression.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DDX41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat