Anti-HTR3B Rabbit Polyclonal Antibody
Catalog # PRSI28-253
Supplier: ProSci Inc.
Specifications
- Antigen name:5-Hydroxytryptamine (serotonin) Receptor 3B
- Antigen symbol:HTR3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:5-HT3B
- Immunogen:A synthetic peptide corresponding a region of human HTR3B.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The product of HTR3B belongs to the ligand-gated ion channel receptor superfamily. HTR3B encodes subunit B of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HTR3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human