Anti-FOXJ2 Rabbit Polyclonal Antibody
Catalog # PRSI27-744
Supplier: ProSci Inc.
Specifications
- Antigen symbol:FOXJ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:FHX
- Immunogen:A synthetic peptide corresponding a region of human FOXJ2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
FOXJ2 is a fork head factor that is expressed in many adult tissues. In the embryo, FOXJ2 expression showed a very early onset during the cleavage stages of preimplantation development. It is capable of activating transcription from promoters containing its sites
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FOXJ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog