Order Entry
Export
ContactUsLinkComponent
Anti-RBPJ Rabbit Polyclonal Antibody
Anti-RBPJ Rabbit Polyclonal Antibody
Catalog # PRSI27-314
Supplier:  ProSci Inc.
undefined
Anti-RBPJ Rabbit Polyclonal Antibody
Catalog # PRSI27-314
Supplier:  ProSci Inc.

Specifications

  • Antigen symbol:
    RBPJ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Dog
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human RBPJ.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

RBPSUH is a DNA binding protein that activates a full transcriptional response but only demonstrates partial anti-apoptotic activity. RTA-mediated redirection of RBPSUH activity from repression to activation is critical for lytic viral replication.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RBPJ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human