Anti-LRRC57 Rabbit Polyclonal Antibody
Catalog # PRSI27-051
Supplier: ProSci Inc.
Specifications
- Antigen name:Leucine Rich Repeat Containing 57
- Antigen symbol:LRRC57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human LRRC57.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The exact function of LRRC57 remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LRRC57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse