Anti-PRSS54 Rabbit Polyclonal Antibody
PRSI26-886
- Antigen name:Protease Serine 54
- Antigen symbol:PRSS54
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human Klkbl4.
- Size:50 µg
- Pk:50 µG
Klkbl4 is a secreted protein. Klkbl4 belongs to the peptidase S1 family, plasma kallikrein subfamily. It contains 1 peptidase S1 domain.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRSS54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human