Order Entry
Export
ContactUsLinkComponent
 
Anti-C20ORF195 Rabbit Polyclonal Antibody
  PRSI26-829
undefined
Anti-C20ORF195 Rabbit Polyclonal Antibody
  PRSI26-829

 

  • Antigen name:
    Chromosome 20 open reading frame 195
  • Antigen symbol:
    C20ORF195
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Chromosome 20 open reading frame 195|CT195_HUMAN|Uncharacterized protein C20orf195.|MGC5356|hypothetical protein LOC79025
  • Immunogen:
    A synthetic peptide corresponding a region of human C20orf195.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

The exact function of C20orf195 remains unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: C20ORF195
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human